4-(三氟甲基)苯甲酸 ≥98.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: α,α,α-三氟对甲苯酸, 对三氟甲基苯甲酸

T1145-25G T1145-5G
TCIAT1145-25GEA 0 CNY
TCIAT1145-25G TCIAT1145-5G
4-(三氟甲基)苯甲酸 ≥98.0% (by titrimetric analysis)
4-(三氟甲基)苯甲酸
分子式: C₈H₅F₃O₂
溶点: 219…220 °C
MDL编号: MFCD00002562
CAS号: 455-24-3
EINECS: 207-242-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ab985f21224bb5b354c8896c920150184565a8e&vid=3ab985f21224bb5b354c8896c920150184565a8e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 221.0 to 225.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR