4-(三氟甲基)苯甲醛 ≥95.0% (by GC)
供应商: TCI
Synonyms:
α,α,α-三氟对甲苯甲醛
T1091-25G
T1091-250G
T1091-5G
TCIAT1091-25GEA
0
CNY
TCIAT1091-25G
TCIAT1091-250G
TCIAT1091-5G
4-(三氟甲基)苯甲醛 ≥95.0% (by GC)
4-(三氟甲基)苯甲醛
分子式:
C₈H₅F₃O 分子量: 174.12 g/mol |
MDL编号:
MFCD00006952 CAS号: 455-19-6 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...