3-甲基吲哚 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 3-甲基吲哚, 粪臭素

M0347-1G M0347-25G M0347-5G
TCIAM0347-1GEA 0 CNY
TCIAM0347-1G TCIAM0347-25G TCIAM0347-5G
3-甲基吲哚 ≥98.0% (by GC)
3-甲基吲哚
分子式: C₉H₉N
分子量: 131.18 g/mol
沸点: 141°C
MDL编号: MFCD00005627
CAS号: 83-34-1
EINECS: 201-471-7
Merck Index: 14,08560
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75dbbf9f8a80be304b13d241c05541519c183458&vid=75dbbf9f8a80be304b13d241c05541519c183458&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 98.0 deg-C
Solubility in EtOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR