异戊酸 ≥99.0% (by GC)

供应商: TCI
Danger

Synonyms: 异丙基乙酸, 3-甲基丁酸

M0182-25ML M0182-500ML
TCIAM0182-25MLEA 0 CNY
TCIAM0182-25ML TCIAM0182-500ML
异戊酸 ≥99.0% (by GC)
异戊酸
分子式: C₅H₁₀O₂
分子量: 102.13 g/mol
沸点: 175°C
溶点: -33
闪点: 79°C
存储条件: Ambient
MDL编号: MFCD00002726
CAS号: 503-74-2
EINECS: 207-975-3
UN号: 2922
ADR: 8 II
Merck Index: 14,05231
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5332f6fcd6a19ddb47f10972fdf65c46398d1a0&vid=a5332f6fcd6a19ddb47f10972fdf65c46398d1a0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR