3-碘-4-甲基苯甲酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
I0707-25G
I0707-5G
TCIAI0707-25GEA
0
CNY
TCIAI0707-25G
TCIAI0707-5G
3-碘-4-甲基苯甲酸 ≥98.0% (by GC, titration analysis)
3-碘-4-甲基苯甲酸
分子式:
C₈H₇IO₂ 分子量: 262.05 g/mol 沸点: 329 |
MDL编号:
MFCD00010392 CAS号: 82998-57-0 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 207.0 to 212.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...