3-氯-2-甲基苯甲酸 ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

C2057-1G C2057-5G
TCIAC2057-1GEA 0 CNY
TCIAC2057-1G TCIAC2057-5G
3-氯-2-甲基苯甲酸 ≥97.0% (by GC, titration analysis)
3-氯-2-甲基苯甲酸
分子式: C₈H₇ClO₂
分子量: 170.6 g/mol
MDL编号: MFCD00053312
CAS号: 7499-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cde78df21ab4cbbd65ec9a2560c5a6d4d62c2e80&vid=cde78df21ab4cbbd65ec9a2560c5a6d4d62c2e80&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR