3-Chloro-2-fluoropyridine ≥98.0% (by GC)

供应商: TCI
Warning

C2515-25G C2515-5G
TCIAC2515-25GEA 0 CNY
TCIAC2515-25G TCIAC2515-5G
3-Chloro-2-fluoropyridine ≥98.0% (by GC)
3-Chloro-2-fluoropyridine
分子式: C₅H₃ClFN
分子量: 131.54 g/mol
沸点: 70°C
闪点: 52°C
MDL编号: MFCD04114144
CAS号: 1480-64-4
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=091184f96ac14654d1ed3d09fabe6bd22d90b67a&vid=091184f96ac14654d1ed3d09fabe6bd22d90b67a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR