3-Amino-4-chloropyridine ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 4-Chloro-3-pyridylamine, 4-Chloropyridin-3-amine

A2395-1G A2395-5G
TCIAA2395-1GEA 0 CNY
TCIAA2395-1G TCIAA2395-5G
3-Amino-4-chloropyridine ≥98.0% (by GC, titration analysis)
3-Amino-4-chloropyridine
分子式: C₅H₅ClN₂
分子量: 128.56 g/mol
存储条件: Refrigerator
MDL编号: MFCD00235124
CAS号: 20511-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02a25ac5748e9c500ac055f0c415439c8a18fa89&vid=02a25ac5748e9c500ac055f0c415439c8a18fa89&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR