3-氨基-1-丙醇 99,0 GC, T

供应商: TCI
Danger

Synonyms: 3-氨基丙醇

A0438-25G A0438-500G
TCIAA0438-25GEA 0 CNY
TCIAA0438-25G TCIAA0438-500G
3-氨基-1-丙醇 99,0 GC, T
3-氨基-1-丙醇
分子式: C₃H₉NO
分子量: 75.11 g/mol
沸点: 188°C
溶点: 11
闪点: 101°C
MDL编号: MFCD00008223
CAS号: 156-87-6
EINECS: 205-864-4
UN号: 2735
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b6c3f5424029e4a4efd18a67ff32aad07703709&vid=7b6c3f5424029e4a4efd18a67ff32aad07703709&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR