Dimidium bromide ≥98.0% (by HPLC)

供应商: TCI
Warning

Synonyms: 2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide

D1815-1G D1815-5G
TCIAD1815-1GEA 0 CNY
TCIAD1815-1G TCIAD1815-5G
Dimidium bromide ≥98.0% (by HPLC)
Dimidium bromide
分子式: C₂₀H₁₈BrN₃
分子量: 380.29 g/mol
存储条件: Ambient
MDL编号: MFCD00011757
CAS号: 518-67-2
EINECS: 208-256-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=524298236bb416a7574b2e12a8bf000d52b317ca&vid=524298236bb416a7574b2e12a8bf000d52b317ca&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR