3,5-吡啶二甲酸 ≥98.0% (通过HPLC,滴定分析)
供应商: TCI
|
Warning
|
P0551-25G
P0551-5G
TCIAP0551-25GEA
0
CNY
TCIAP0551-25G
TCIAP0551-5G
3,5-吡啶二甲酸 ≥98.0% (通过HPLC,滴定分析)
3,5-吡啶二甲酸
分子式:
C₇H₅NO₄ 沸点: 357 °C (1013 hPa) 溶点: 320…325 °C 存储条件: Ambient |
MDL编号:
MFCD00006393 CAS号: 499-81-0 EINECS: 207-893-8 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in N,N-DMF | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...