3,5-二甲氧基苯甲酰氯 ≥98.0% (通过GC,滴定分析)

供应商: TCI
Danger

D2609-25G D2609-5G
TCIAD2609-25GEA 0 CNY
TCIAD2609-25G TCIAD2609-5G
3,5-二甲氧基苯甲酰氯 ≥98.0% (通过GC,滴定分析)
3,5-二甲氧基苯甲酰氯
分子式: C₉H₉ClO₃
沸点: 284 °C (1013 hPa)
溶点: 35…36 °C
MDL编号: MFCD00000676
CAS号: 17213-57-9
EINECS: 241-256-5
UN号: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eed740e1732c8ef57783e4a7db7269cc2ed61a3&vid=8eed740e1732c8ef57783e4a7db7269cc2ed61a3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR