2-丙基戊酸 ≥99.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 丙戊酸

P0823-25ML P0823-500ML
TCIAP0823-25MLEA 0 CNY
TCIAP0823-25ML TCIAP0823-500ML
2-丙基戊酸 ≥99.0% (by GC, titration analysis)
2-丙基戊酸
分子式: C₈H₁₆O₂
沸点: 220°C
密度: 0,905 g/cm³ (20 °C)
闪点: 126°C
存储条件: Ambient
MDL编号: MFCD00002672
CAS号: 99-66-1
EINECS: 202-777-3
ADR: 8,III
Merck Index: 14,09913
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7414c95fa80bcdee9d1429fd6908a2ad0c536a2&vid=d7414c95fa80bcdee9d1429fd6908a2ad0c536a2&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR