2-甲基咪唑 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 2-甲基-1,3-氮杂茂

M0345-25G M0345-500G
TCIAM0345-25GEA 0 CNY
TCIAM0345-25G TCIAM0345-500G
2-甲基咪唑 ≥98.0% (by GC, titration analysis)
2-甲基咪唑
分子式: C₄H₆N₂
分子量: 82.11 g/mol
沸点: 267
闪点: 155
存储条件: Ambient
MDL编号: MFCD00005190
CAS号: 693-98-1
EINECS: 211-765-7
UN号: 3263
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6fdc98018da9047d0e67089ec2e54fc4c109446&vid=c6fdc98018da9047d0e67089ec2e54fc4c109446&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 144.0 to 148.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR