2-甲基己酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
2-甲己酸
M0947-25ML
M0947-5ML
TCIAM0947-25MLEA
0
CNY
TCIAM0947-25ML
TCIAM0947-5ML
2-甲基己酸 ≥98.0% (by GC, titration analysis)
2-甲基己酸
分子式:
C₇H₁₄O₂ 分子量: 130.19 g/mol 沸点: 72°C 密度: 0.929 闪点: 105°C 存储条件: Ambient |
MDL编号:
MFCD00002674 CAS号: 4536-23-6 EINECS: 224-883-9 UN号: 3265 ADR: 8 III |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...