2-甲基-1-丙醇 ≥99.0% (by GC)
供应商: TCI
Danger
Synonyms: 异丁醇, 异丙基甲醇
I0094-25ML I0094-500ML
TCIAI0094-25MLEA 0 CNY
TCIAI0094-25ML TCIAI0094-500ML
2-甲基-1-丙醇 ≥99.0% (by GC)
2-甲基-1-丙醇
分子式: (CH₃)₂CHCH₂OH
分子量: 74.12 g/mol
沸点: 108°C
溶点: –108°C
闪点: 30°C
存储条件: Ambient
MDL编号: MFCD00004740
CAS号: 78-83-1
EINECS: 201-148-0
UN号: 1212
ADR: 3 III
Merck Index: 14,05131
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ecd78fb5557bd355bac0babd431a25bc058e70d6&vid=ecd78fb5557bd355bac0babd431a25bc058e70d6&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %