2-甲氧基萘 ≥98.0% (by GC)
供应商: TCI
Synonyms:
2-萘甲醚
M0117-25G
M0117-500G
TCIAM0117-25GEA
0
CNY
TCIAM0117-25G
TCIAM0117-500G
2-甲氧基萘 ≥98.0% (by GC)
2-甲氧基萘
分子式:
C₁₁H₁₀O 分子量: 158.2 g/mol 沸点: 274 |
MDL编号:
MFCD00004061 CAS号: 93-04-9 EINECS: 202-213-6 Merck Index: 14,05997 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 72.0 to 76.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...