异硫氰酸异丙酯 ≥98.0% (通过GC)
供应商: TCI
|
Danger
|
I0520-25ML
I0520-5ML
TCIAI0520-25MLEA
0
CNY
TCIAI0520-25ML
TCIAI0520-5ML
异硫氰酸异丙酯 ≥98.0% (通过GC)
异硫氰酸异丙酯
分子式:
C₄H₇NS 沸点: 138°C 密度: 0,9475 g/cm³ (20 °C) |
MDL编号:
MFCD00037830 CAS号: 2253-73-8 EINECS: 218-851-3 UN号: 1992 ADR: 3,II |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...