邻氟苯甲酰氯 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: o-Fluorobenzoyl chloride

F0195-25G F0195-500G
TCIAF0195-25GEA 0 CNY
TCIAF0195-25G TCIAF0195-500G
邻氟苯甲酰氯 ≥98.0% (by GC, titration analysis)
邻氟苯甲酰氯
分子式: C₇H₄ClFO
分子量: 158.56 g/mol
沸点: 206
闪点: 82°C
存储条件: Ambient
MDL编号: MFCD00000656
CAS号: 393-52-2
EINECS: 206-887-2
UN号: 3265
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e31175a4a574e00c34f80bcc90aeebadcc7cd544&vid=e31175a4a574e00c34f80bcc90aeebadcc7cd544&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR