2-氟-6-甲基吡啶 ≥95.0% (by GC)

供应商: TCI
Warning

Synonyms: 6-氟-2-甲基吡啶

F0520-1G F0520-5G
TCIAF0520-1GEA 0 CNY
TCIAF0520-1G TCIAF0520-5G
2-氟-6-甲基吡啶 ≥95.0% (by GC)
2-氟-6-甲基吡啶
分子式: C₆H₆FN
分子量: 111.12 g/mol
沸点: 142°C
闪点: 66°C
MDL编号: MFCD00041226
CAS号: 407-22-7
EINECS: 206-980-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04acec02b9963aff6f5e4f684da26e82b82ff1df&vid=04acec02b9963aff6f5e4f684da26e82b82ff1df&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR