2-氟-5-甲基吡啶 ≥98.0% (通过GC)
供应商: TCI
|
Warning
|
F0581-1G
F0581-5G
TCIAF0581-1GEA
0
CNY
TCIAF0581-1G
TCIAF0581-5G
2-氟-5-甲基吡啶 ≥98.0% (通过GC)
2-氟-5-甲基吡啶
分子式:
C₆H₆FN 沸点: 155°C 密度: 1,077 g/cm³ (20 °C) 闪点: 48°C |
MDL编号:
MFCD00041225 CAS号: 2369-19-9 UN号: 1993 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...