2-乙基咪唑 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 2-Ethyl-1H-imidazole

E0233-25G E0233-500G
TCIAE0233-25GEA 0 CNY
TCIAE0233-25G TCIAE0233-500G
2-乙基咪唑 ≥98.0% (by GC, titration analysis)
2-乙基咪唑
分子式: C₅H₈N₂
分子量: 96.13 g/mol
沸点: 268
存储条件: Ambient
MDL编号: MFCD00005192
CAS号: 1072-62-4
EINECS: 214-011-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a367b2e4152d4ad14aaef6802146a98d98abf447&vid=a367b2e4152d4ad14aaef6802146a98d98abf447&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 88.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR