2-乙基己酸 ≥99.0% (通过GC)

供应商: TCI
Danger

E0120-25ML E0120-500ML
TCIAE0120-25MLEA 0 CNY
TCIAE0120-25ML TCIAE0120-500ML
2-乙基己酸 ≥99.0% (通过GC)
2-乙基己酸
分子式: C₈H₁₆O₂
沸点: 228°C
密度: 0,91 g/cm³ (20 °C)
闪点: 120°C
存储条件: Ambient
MDL编号: MFCD00002675
CAS号: 149-57-5
EINECS: 205-743-6
UN号: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01e32d8af2c10f46a61bcd5f310cc6f10ff96d10&vid=01e32d8af2c10f46a61bcd5f310cc6f10ff96d10&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR