2-氯吡嗪 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: Chloropyrazine

C1093-25G C1093-5G
TCIAC1093-25GEA 0 CNY
TCIAC1093-25G TCIAC1093-5G
2-氯吡嗪 ≥98.0% (by GC)
2-氯吡嗪
分子式: C₄H₃ClN₂
分子量: 114.53 g/mol
沸点: 93°C
闪点: 57°C
存储条件: Ambient
MDL编号: MFCD00006124
CAS号: 14508-49-7
EINECS: 238-517-0
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae26bbe8734fa62cc38c351b86f107f147b4fbf9&vid=ae26bbe8734fa62cc38c351b86f107f147b4fbf9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR