2-Chlorohexane ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-Hexyl chloride

C0890-25G C0890-5G
TCIAC0890-25GEA 0 CNY
TCIAC0890-25G TCIAC0890-5G
2-Chlorohexane ≥98.0% (by GC)
2-Chlorohexane
分子式: C₆H₁₃Cl
分子量: 120.62 g/mol
沸点: 122°C
存储条件: Refrigerator
MDL编号: MFCD00039351
CAS号: 638-28-8
EINECS: 211-329-6
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b560ab5888b8d18a65b70f6e1a5c82eb69026cd1&vid=b560ab5888b8d18a65b70f6e1a5c82eb69026cd1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR