2-溴-5-氟吡啶 ≥98.0% (by GC)

供应商: TCI
Warning

B2799-25G B2799-5G
TCIAB2799-25GEA 0 CNY
TCIAB2799-25G TCIAB2799-5G
2-溴-5-氟吡啶 ≥98.0% (by GC)
2-溴-5-氟吡啶
分子式: C₅H₃BrFN
分子量: 175.99 g/mol
沸点: 83°C
MDL编号: MFCD00234011
CAS号: 41404-58-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b274c4c33e7dbe3ab193bd59dea2b9360a61e36f&vid=b274c4c33e7dbe3ab193bd59dea2b9360a61e36f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR