2-Amino-6-methyl-3-nitropyridine ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
6-Methyl-3-nitro-2-pyridylamine, 6-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-6-picoline, 2-Amino-3-nitro-6-picoline
A2334-1G
A2334-5G
TCIAA2334-1GEA
0
CNY
TCIAA2334-1G
TCIAA2334-5G
2-Amino-6-methyl-3-nitropyridine ≥98.0% (by GC)
2-Amino-6-methyl-3-nitropyridine
分子式:
C₆H₇N₃O₂ 分子量: 153.14 g/mol |
MDL编号:
MFCD00047443 CAS号: 21901-29-1 |
|
产品技术规格
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 154.0 to 158.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...