2-氨基-5-碘吡啶 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 5-Iodopyridin-2-amine, 5-Iodo-2-pyridylamine

A1842-1G A1842-25G A1842-5G
TCIAA1842-1GEA 0 CNY
TCIAA1842-1G TCIAA1842-25G TCIAA1842-5G
2-氨基-5-碘吡啶 ≥98.0% (by GC, titration analysis)
2-氨基-5-碘吡啶
分子式: C₅H₅IN₂
分子量: 220.01 g/mol
沸点: 271
存储条件: Refrigerator
MDL编号: MFCD00160312
CAS号: 20511-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8496f989134c7767c6d40adb89cadbc59d0a64ad&vid=8496f989134c7767c6d40adb89cadbc59d0a64ad&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 127.0 to 132.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR