2-氨基-1-丁醇 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
2-Aminobutan-1-ol, (±)-2-Amino-1-butanol
A0684-25G
A0684-500G
TCIAA0684-25GEA
0
CNY
TCIAA0684-25G
TCIAA0684-500G
2-氨基-1-丁醇 ≥98.0% (by GC, titration analysis)
2-氨基-1-丁醇
分子式:
C₄H₁₁NO 分子量: 89.14 g/mol 沸点: 180°C 溶点: -2 闪点: 84°C 存储条件: Ambient |
MDL编号:
MFCD00008095 CAS号: 96-20-8 EINECS: 202-488-2 UN号: 3267 ADR: 8 III Merck Index: 14,00427 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...