2,6-二氯苯甲酰氯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 2,6-Dichlorobenzoyl Chloride

D2064-25G D2064-500G
TCIAD2064-25GEA 0 CNY
TCIAD2064-25G TCIAD2064-500G
2,6-二氯苯甲酰氯 ≥98.0% (by GC)
2,6-二氯苯甲酰氯
分子式: C₇H₃Cl₃O
分子量: 209.46 g/mol
沸点: 127°C
溶点: 18
闪点: 147°C
存储条件: Ambient
MDL编号: MFCD00000662
CAS号: 4659-45-4
EINECS: 225-102-4
UN号: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4feb89aaa3979701564b7d4b733941aec51a03ba&vid=4feb89aaa3979701564b7d4b733941aec51a03ba&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR