2,6-Di-tert-butylnaphthalene ≥98.0% (by GC)

供应商: TCI

D2369-5G D2369-25G
TCIAD2369-5GEA 0 CNY
TCIAD2369-5G TCIAD2369-25G
2,6-Di-tert-butylnaphthalene ≥98.0% (by GC)
2,6-Di-tert-butylnaphthalene
分子式: C₁₈H₂₄
MDL编号: MFCD00004110
CAS号: 3905-64-4
EINECS: 223-461-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=881c5b109d7a4ea73051c785a711e096a582fe12&vid=881c5b109d7a4ea73051c785a711e096a582fe12&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR