2,4-二氯嘧啶 ≥98.0% (by GC)

供应商: TCI
Warning

D2310-25G D2310-5G
TCIAD2310-25GEA 0 CNY
TCIAD2310-25G TCIAD2310-5G
2,4-二氯嘧啶 ≥98.0% (by GC)
2,4-二氯嘧啶
分子式: C₄H₂Cl₂N₂
沸点: 101°C
溶点: 61 °C
MDL编号: MFCD00006061
CAS号: 3934-20-1
EINECS: 223-508-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19f72e9417fbe7aec4dcfe8b55985780b54b379f&vid=19f72e9417fbe7aec4dcfe8b55985780b54b379f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 59.0 to 62.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR