2,3,6-三氟苯甲醛 ≥98.0% (by GC)

供应商: TCI
Warning

T2318-1G T2318-5G
TCIAT2318-1GEA 0 CNY
TCIAT2318-1G TCIAT2318-5G
2,3,6-三氟苯甲醛 ≥98.0% (by GC)
2,3,6-三氟苯甲醛
分子式: C₇H₃F₃O
闪点: 76°C
存储条件: Refrigerator
MDL编号: MFCD00061195
CAS号: 104451-70-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f201dcf23284f41431bef4697b3ddc8ece961332&vid=f201dcf23284f41431bef4697b3ddc8ece961332&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR