o-Propyltoluene ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 2-Propyltoluene, 1-Methyl-2-propylbenzene

P0747-25ML P0747-5ML
TCIAP0747-25MLEA 0 CNY
TCIAP0747-25ML TCIAP0747-5ML
o-Propyltoluene ≥98.0% (by GC)
o-Propyltoluene
分子式: C₁₀H₁₄
沸点: 185°C
MDL编号: MFCD00059246
CAS号: 1074-17-5
EINECS: 214-037-7
UN号: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93cb8da6cd3c65627429bad519ae048937199066&vid=93cb8da6cd3c65627429bad519ae048937199066&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR