1-甲氧基萘 ≥98.0% (by GC)
供应商: TCI
|
|
Synonyms:
1-萘甲醚
M0116-25G
M0116-500G
TCIAM0116-25GEA
0
CNY
TCIAM0116-25G
TCIAM0116-500G
1-甲氧基萘 ≥98.0% (by GC)
1-甲氧基萘
分子式:
C₁₁H₁₀O 沸点: 271°C 溶点: 6°C 密度: 1,09 g/cm³ (20 °C) |
MDL编号:
MFCD00003924 CAS号: 2216-69-5 EINECS: 218-696-1 UN号: 3082 ADR: 9,III |
|
产品技术规格
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...