1,1,1-三氟-3-碘丙烷 ≥98.0% (by GC)

供应商: TCI

Synonyms: 1,1,1-Trifluoro-3-iodopropane, 3-Iodo-1,1,1-trifluoropropane

T2588-25G T2588-5G
TCIAT2588-25GEA 0 CNY
TCIAT2588-25G TCIAT2588-5G
1,1,1-三氟-3-碘丙烷 ≥98.0% (by GC)
1,1,1-三氟-3-碘丙烷
分子式: C₃H₄F₃I
沸点: 90°C
MDL编号: MFCD00038531
CAS号: 460-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcc59713bab6d6575d6ecf5b5913a3d823e06d6b&vid=fcc59713bab6d6575d6ecf5b5913a3d823e06d6b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR