4-羟基二苯甲酮 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: (4-Hydroxyphenyl)(phenyl)methanone, p-Hydroxybenzophenone

H0222-25G H0222-500G
TCIAH0222-25GEA 0 CNY
TCIAH0222-25G TCIAH0222-500G
4-羟基二苯甲酮 ≥98.0% (by GC, titration analysis)
4-羟基二苯甲酮
分子式: C₁₃H₁₀O₂
分子量: 198.22 g/mol
沸点: 367
存储条件: Ambient
MDL编号: MFCD00002355
CAS号: 1137-42-4
EINECS: 214-507-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=398412ebc56c5a5947bf9e4491ec3b88d9b7771c&vid=398412ebc56c5a5947bf9e4491ec3b88d9b7771c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 133.0 to 136.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR