3-甲基-2-硝基苯酚 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-羟基-2-硝基甲苯, 2-硝基间甲酚

N0552-25G N0552-5G
TCIAN0552-25GEA 0 CNY
TCIAN0552-25G TCIAN0552-5G
3-甲基-2-硝基苯酚 ≥98.0% (by GC, titration analysis)
3-甲基-2-硝基苯酚
分子式: C₇H₇NO₃
分子量: 153.14 g/mol
沸点: 108°C
存储条件: Refrigerator
MDL编号: MFCD00007125
CAS号: 4920-77-8
EINECS: 225-546-9
UN号: 2446
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c4509680a6c5a485f8a21cbc9ea0ea739e6a093&vid=3c4509680a6c5a485f8a21cbc9ea0ea739e6a093&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 38.0 to 41.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR