4-氟-2-硝基苯甲醚 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-Fluoro-1-methoxy-2-nitrobenzene

F0615-25G F0615-5G
TCIAF0615-25GEA 0 CNY
TCIAF0615-25G TCIAF0615-5G
4-氟-2-硝基苯甲醚 ≥98.0% (by GC)
4-氟-2-硝基苯甲醚
分子式: C₇H₆FNO₃
分子量: 171.13 g/mol
存储条件: Ambient
MDL编号: MFCD00013375
CAS号: 445-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56b0c20649aa5b8dcda4162f8733cb2e2d557035&vid=56b0c20649aa5b8dcda4162f8733cb2e2d557035&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR