4-氟苯基异硫氰酸酯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 1-Fluoro-4-isothiocyanatobenzene

F0644-25G F0644-5G
TCIAF0644-25GEA 0 CNY
TCIAF0644-25G TCIAF0644-5G
4-氟苯基异硫氰酸酯 ≥98.0% (by GC)
4-氟苯基异硫氰酸酯
分子式: C₇H₄FNS
分子量: 153.18 g/mol
沸点: 228°C
闪点: 89°C
存储条件: Refrigerator
MDL编号: MFCD00004809
CAS号: 1544-68-9
EINECS: 216-280-4
UN号: 2922
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c0d002295a7223dea193b728da7c226c15857f6&vid=9c0d002295a7223dea193b728da7c226c15857f6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Green clear liquid to slightly cloudy liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR