4-氟二苯甲酮 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
F0161-25G
F0161-5G
TCIAF0161-25GEA
0
CNY
TCIAF0161-25G
TCIAF0161-5G
4-氟二苯甲酮 ≥99.0% (by GC)
4-氟二苯甲酮
分子式:
C₁₃H₉FO 分子量: 200.21 g/mol 沸点: 161°C 溶点: 49 |
MDL编号:
MFCD00000352 CAS号: 345-83-5 EINECS: 206-463-7 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Solubility in Methanol | almost transparency |
Melting point | 47.0 to 50.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...