3′,4′,5′-三甲氧基苯乙酮 ≥98.0% (通过GC)

供应商: TCI

T0898-25G T0898-5G
TCIAT0898-25GEA 0 CNY
TCIAT0898-25G TCIAT0898-5G
3′,4′,5′-三甲氧基苯乙酮 ≥98.0% (通过GC)
3′,4′,5′-三甲氧基苯乙酮
分子式: C₁₁H₁₄O₄
沸点: 224°C
溶点: 73…74 °C
MDL编号: MFCD00008738
CAS号: 1136-86-3
EINECS: 214-501-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e7ffe5a3e42443db1b261ce06b91f41278f772e&vid=1e7ffe5a3e42443db1b261ce06b91f41278f772e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light red to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 83.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR