2',4',6'-Trifluoroacetophenone ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1-(2,4,6-Trifluorophenyl)ethanone, 2,4,6-Trifluoroacetophenone, 1-(2,4,6-Trifluorophenyl)ethan-1-one
T2733-25G
T2733-5G
TCIAT2733-25GEA
0
CNY
TCIAT2733-25G
TCIAT2733-5G
2',4',6'-Trifluoroacetophenone ≥98.0% (by GC)
2',4',6'-Trifluoroacetophenone
分子式:
C₈H₅F₃O 分子量: 174.12 g/mol 沸点: 190°C |
MDL编号:
MFCD00061194 CAS号: 51788-77-3 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...