2′,3′,4′,5′,6′-五氟苯乙酮 ≥98.0% (by GC)
供应商: TCI
Warning
P0921-25G P0921-5G
TCIAP0921-25GEA 0 CNY
TCIAP0921-25G TCIAP0921-5G
2′,3′,4′,5′,6′-五氟苯乙酮 ≥98.0% (by GC)
2′,3′,4′,5′,6′-五氟苯乙酮
分子式: C₈H₃F₅O
沸点: 131°C
密度: 1,47 g/cm³ (20 °C)
闪点: 66°C
MDL编号: MFCD00000296
CAS号: 652-29-9
EINECS: 211-487-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc4fc0248302c0e701e9b1a3c9434947a5f2fa2f&vid=fc4fc0248302c0e701e9b1a3c9434947a5f2fa2f&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %