1-Butyl-2-nitrobenzene ≥96.0% (by GC)

供应商: TCI

B3091-25G B3091-5G
TCIAB3091-25GEA 0 CNY
TCIAB3091-25G TCIAB3091-5G
1-Butyl-2-nitrobenzene ≥96.0% (by GC)
1-Butyl-2-nitrobenzene
分子式: C₁₀H₁₃NO₂
分子量: 179.22 g/mol
沸点: 127°C
CAS号: 7137-55-5
EINECS: 230-435-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b1045b5df3d276f8159d35072ef74919fd70a63&vid=0b1045b5df3d276f8159d35072ef74919fd70a63&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR