2-溴-5-硝基三氟甲苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Bromo-5-nitrobenzotrifluoride, 1-Bromo-4-nitro-2-(trifluoromethyl)benzene
B3686-25G
B3686-5G
TCIAB3686-25GEA
0
CNY
TCIAB3686-25G
TCIAB3686-5G
2-溴-5-硝基三氟甲苯 ≥98.0% (by GC)
2-溴-5-硝基三氟甲苯
分子式:
C₇H₃BrF₃NO₂ 分子量: 270.01 g/mol 沸点: 88°C |
MDL编号:
MFCD00014707 CAS号: 367-67-9 |
|
产品技术规格
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...