2-溴-5-氟三氟甲苯 ≥97.0% (by GC)

供应商: TCI
Warning

B2800-25G B2800-5G
TCIAB2800-25GEA 0 CNY
TCIAB2800-25G TCIAB2800-5G
2-溴-5-氟三氟甲苯 ≥97.0% (by GC)
2-溴-5-氟三氟甲苯
分子式: C₇H₃BrF₄
分子量: 243 g/mol
沸点: 143°C
闪点: 64°C
MDL编号: MFCD00040937
CAS号: 40161-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=50e95359141e693deba5879cfaf679c8a1eb145c&vid=50e95359141e693deba5879cfaf679c8a1eb145c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR