2-Bromo-4-chloroanisole ≥98.0% (by GC)

供应商: TCI

Synonyms: 2-Bromo-4-chloro-1-methoxybenzene

B3111-25G B3111-5G
TCIAB3111-25GEA 0 CNY
TCIAB3111-25G TCIAB3111-5G
2-Bromo-4-chloroanisole ≥98.0% (by GC)
2-Bromo-4-chloroanisole
分子式: C₇H₆BrClO
分子量: 221.48 g/mol
沸点: 127°C
MDL编号: MFCD00079705
CAS号: 60633-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d0d910ad22c8c6e1d111d854dae8e79cc57469b&vid=1d0d910ad22c8c6e1d111d854dae8e79cc57469b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR