3-溴-4-氟三氟甲苯 ≥96.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2-Bromo-1-fluoro-4-(trifluoromethyl)benzene, 3-Bromo-4-fluorobenzotrifluoride, 1-Bromo-2-fluoro-5-(trifluoromethyl)benzene
B1421-25G
B1421-5G
TCIAB1421-25GEA
0
CNY
TCIAB1421-25G
TCIAB1421-5G
3-溴-4-氟三氟甲苯 ≥96.0% (by GC)
3-溴-4-氟三氟甲苯
分子式:
C₇H₃BrF₄ 分子量: 243 g/mol 沸点: 60°C 存储条件: Refrigerator |
MDL编号:
MFCD00040938 CAS号: 68322-84-9 |
|
产品技术规格
Purity (GC) | min. 96.0 % |
Appearance | Colorless to Light orange to Yellow clear liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...