1-溴-2-(三氟甲氧基)苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
T2147-25G
T2147-5G
TCIAT2147-25GEA
0
CNY
TCIAT2147-25G
TCIAT2147-5G
1-溴-2-(三氟甲氧基)苯 ≥98.0% (by GC)
1-溴-2-(三氟甲氧基)苯
分子式:
C₇H₄BrF₃O 分子量: 241.01 g/mol 闪点: 60°C |
MDL编号:
MFCD00040942 CAS号: 64115-88-4 UN号: 1993 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...